Assay Details
| Gene Symbol: tmem106b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016812.2 | NP_001016812.1 | 1 - 2 | 233 | 66 | |
| GenBank mRNA | BC154062.1 | - | 2 - 3 | 252 | 66 | |
| CR855566.2 | - | 1 - 2 | 233 | 66 | ||
More Information
Set Membership: |
|

tmem106b
Genomic Map