Assay Details
Gene Symbol: nob1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016830.2 | NP_001016830.1 | 2 - 3 | 215 | 71 | |
XM_012961142.2 | XP_012816596.1 | 2 - 3 | 478 | 71 | ||
GenBank mRNA | CR855508.2 | - | 2 - 3 | 215 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
