Assay Details
| Gene Symbol: tmx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016906.2 | NP_001016906.1 | 7 - 8 | 1007 | 66 | |
| XM_012968448.2 | XP_012823902.1 | 7 - 8 | 1094 | 66 | ||
| GenBank mRNA | CR848507.2 | - | 7 - 8 | 1007 | 66 | |
More Information
Set Membership: |
|

tmx1
Genomic Map