Assay Details
Gene Symbol: gcm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016915.2 | NP_001016915.1 | 3 - 4 | 467 | 62 | |
GenBank mRNA | CR848297.2 | - | 3 - 4 | 467 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
