Assay Details
Gene Symbol: hnrnpa2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016926.3 | NP_001016926.2 | 2 - 3 | 235 | 88 | |
NM_001205342.1 | NP_001192271.1 | 2 - 3 | 235 | 88 | ||
XM_018094667.1 | XP_017950156.1 | 2 - 3 | 434 | 88 | ||
XM_018094669.1 | XP_017950158.1 | 2 - 3 | 434 | 88 | ||
GenBank mRNA | BC135350.1 | - | 2 - 3 | 202 | 88 | |
CR762263.2 | - | 2 - 3 | 143 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
