Assay Details
| Gene Symbol: med26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016996.2 | NP_001016996.1 | 2 - 3 | 293 | 98 | |
| XM_018093627.1 | XP_017949116.1 | 2 - 3 | 432 | 98 | ||
| GenBank mRNA | CR848161.2 | - | 2 - 3 | 293 | 98 | |
More Information
Set Membership: |
|

med26
Genomic Map