Assay Details
| Gene Symbol: wnt8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017208.2 | NP_001017208.1 | 5 - 6 | 547 | 75 | |
| GenBank mRNA | CR760475.2 | - | 5 - 6 | 547 | 75 | |
More Information
Set Membership: |
|

wnt8a
Genomic Map