Assay Details
| Gene Symbol: exosc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037265.1 | NP_001032342.1 | 4 - 5 | 436 | 66 | |
| GenBank mRNA | BC088546.1 | - | 4 - 5 | 417 | 66 | |
| BC121333.1 | - | 4 - 5 | 436 | 66 | ||
| CR759986.2 | - | 4 - 5 | 436 | 66 | ||
More Information
Set Membership: |
|

exosc7
Genomic Map