Assay Details
| Gene Symbol: hebp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039739.1 | NP_001034828.1 | 2 - 3 | 308 | 90 | |
| GenBank mRNA | BC089188.1 | - | 2 - 3 | 275 | 90 | |
| BC118708.1 | - | 2 - 3 | 277 | 90 | ||
| BC158372.1 | - | 2 - 3 | 280 | 90 | ||
| CR760976.2 | - | 2 - 3 | 308 | 90 | ||
More Information
Set Membership: |
|

hebp2
Genomic Map