Assay Details
| Gene Symbol: yipf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015932.3 | NP_001015932.1 | 4 - 5 | 300 | 77 | |
| XM_018096248.1 | XP_017951737.1 | 3 - 4 | 269 | 77 | ||
| GenBank mRNA | BC122940.1 | - | 4 - 5 | 300 | 77 | |
| CR926375.2 | - | 4 - 5 | 296 | 77 | ||
More Information
Set Membership: |
|

yipf6
Genomic Map