Assay Details
| Gene Symbol: zdhhc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044406.1 | NP_001037871.1 | 3 - 4 | 471 | 63 | |
| XM_018093887.1 | XP_017949376.1 | 3 - 4 | 492 | 63 | ||
| XM_018093888.1 | XP_017949377.1 | 3 - 4 | 357 | 63 | ||
| GenBank mRNA | AY871209.1 | - | 3 - 4 | 471 | 63 | |
| CU075592.1 | - | 3 - 4 | 474 | 63 | ||
More Information
Set Membership: |
|

zdhhc8
Genomic Map