Assay Details
| Gene Symbol: slc38a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044435.1 | NP_001037900.1 | 8 - 9 | 1427 | 58 | |
| XM_012961231.2 | XP_012816685.1 | 9 - 10 | 1228 | 58 | ||
| XM_012961232.2 | XP_012816686.1 | 9 - 10 | 1277 | 58 | ||
| XM_018093079.1 | XP_017948568.1 | 9 - 10 | 1240 | 58 | ||
| GenBank mRNA | CR760593.2 | - | 8 - 9 | 1427 | 58 | |
More Information
Set Membership: |
|

slc38a8
Genomic Map