Assay Details
| Gene Symbol: glis1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044465.1 | NP_001037930.1 | 6 - 7 | 673 | 82 | |
| XM_018093081.1 | XP_017948570.1 | 8 - 9 | 1915 | 82 | ||
| GenBank mRNA | CR761307.2 | - | 6 - 7 | 673 | 82 | |
More Information
Set Membership: |
|

glis1
Genomic Map