Assay Details
| Gene Symbol: slc7a6os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044467.1 | NP_001037932.1 | 3 - 4 | 748 | 108 | |
| XM_012961243.2 | XP_012816697.1 | 3 - 4 | 781 | 108 | ||
| XM_012961244.2 | XP_012816698.1 | 3 - 4 | 781 | 108 | ||
| XM_018093082.1 | XP_017948571.1 | 3 - 4 | 727 | 108 | ||
| XM_018093083.1 | XP_017948572.1 | 3 - 4 | 781 | 108 | ||
| GenBank mRNA | CR761340.2 | - | 3 - 4 | 748 | 108 | |
More Information
Set Membership: |
|

slc7a6os
Genomic Map