Assay Details
| Gene Symbol: map1lc3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044522.1 | NP_001037987.1 | 3 - 4 | 236 | 83 | |
| GenBank mRNA | BC155037.1 | - | 3 - 4 | 279 | 83 | |
| CR762033.2 | - | 3 - 4 | 236 | 83 | ||
More Information
Set Membership: |
|

map1lc3c
Genomic Map