Assay Details
| Gene Symbol: usp44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078921.1 | NP_001072389.1 | 4 - 5 | 1884 | 88 | |
| XM_012968613.2 | XP_012824067.1 | 4 - 5 | 1997 | 88 | ||
| XM_012968614.2 | XP_012824068.1 | 4 - 5 | 1956 | 88 | ||
| GenBank mRNA | BC121579.1 | - | 4 - 5 | 1884 | 88 | |
| CU075840.1 | - | 4 - 5 | 1926 | 88 | ||
More Information
Set Membership: |
|

usp44
Genomic Map