Assay Details
| Gene Symbol: gpx7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078936.1 | NP_001072404.1 | 1 - 2 | 136 | 68 | |
| GenBank mRNA | BC121615.1 | - | 1 - 2 | 136 | 68 | |
| CU075482.1 | - | 1 - 2 | 68 | |||
More Information
Set Membership: |
|

gpx7
Genomic Map