Assay Details
| Gene Symbol: tmem14c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078956.1 | NP_001072424.1 | 3 - 4 | 365 | 77 | |
| GenBank mRNA | BC121665.1 | - | 3 - 4 | 365 | 77 | |
| BC135365.1 | - | 3 - 4 | 238 | 77 | ||
More Information
Set Membership: |
|

tmem14c
Genomic Map