Assay Details
| Gene Symbol: mcur1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079377.1 | NP_001072845.1 | 8 - 9 | 969 | 78 | |
| XM_018094709.1 | XP_017950198.1 | 8 - 9 | 1062 | 78 | ||
| XM_018094710.1 | XP_017950199.1 | 7 - 8 | 994 | 78 | ||
| GenBank mRNA | BC122047.1 | - | 8 - 9 | 969 | 78 | |
More Information
Set Membership: |
|

mcur1
Genomic Map