Assay Details
| Gene Symbol: tmem19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016079.3 | NP_001016079.1 | 4 - 5 | 560 | 77 | |
| XM_012958991.2 | XP_012814445.1 | 4 - 5 | 632 | 77 | ||
| XM_018092069.1 | XP_017947558.1 | 4 - 5 | 646 | 77 | ||
| GenBank mRNA | BC121999.1 | - | 4 - 5 | 560 | 77 | |
| CR762219.2 | - | 4 - 5 | 480 | 77 | ||
More Information
Set Membership: |
|

tmem19
Genomic Map