Assay Details
| Gene Symbol: tmem38b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016127.3 | NP_001016127.1 | 2 - 3 | 286 | 107 | |
| GenBank mRNA | BC135582.1 | - | 2 - 3 | 286 | 107 | |
| CR762060.2 | - | 2 - 3 | 279 | 107 | ||
More Information
Set Membership: |
|

tmem38b
Genomic Map