Assay Details
| Gene Symbol: tspan31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017068.3 | NP_001017068.1 | 5 - 6 | 627 | 73 | |
| XM_012957025.2 | XP_012812479.1 | 5 - 6 | 740 | 73 | ||
| GenBank mRNA | BC135652.1 | - | 5 - 6 | 627 | 73 | |
| CR761572.2 | - | 5 - 6 | 617 | 73 | ||
More Information
Set Membership: |
|

tspan31
Genomic Map