Assay Details
| Gene Symbol: spcs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017094.3 | NP_001017094.2 | 208 | 73 | ||
| GenBank mRNA | AL858171.2 | - | 1 - 2 | 151 | 73 | |
| BC135349.1 | - | 1 - 2 | 222 | 73 | ||
| CR585316.1 | - | 1 - 2 | 221 | 73 | ||
| EL832300.1 | - | 1 - 2 | 346 | 73 | ||
More Information
Set Membership: |
|

spcs3
Genomic Map