Assay Details
Gene Symbol: npy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079062.1 | NP_001072530.1 | 2 - 3 | 251 | 104 | |
XM_012964911.2 | XP_012820365.1 | 3 - 4 | 519 | 104 | ||
GenBank mRNA | BC121985.1 | - | 2 - 3 | 251 | 104 | |
BC135387.1 | - | 2 - 3 | 304 | 104 | ||
KT202317.1 | - | 2 - 3 | 304 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
