Assay Details
| Gene Symbol: rilpl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079078.2 | NP_001072546.2 | 3 - 4 | 699 | 75 | |
| XM_012965440.2 | XP_012820894.1 | 3 - 4 | 572 | 75 | ||
| GenBank mRNA | BC122050.1 | - | 3 - 4 | 568 | 75 | |
| CU075390.1 | - | 3 - 4 | 647 | 75 | ||
| CX931705.1 | - | 3 - 4 | 699 | 75 | ||
More Information
Set Membership: |
|

rilpl2
Genomic Map