Assay Details
| Gene Symbol: fam222a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079118.1 | NP_001072586.1 | 2 - 3 | 1153 | 97 | |
| GenBank mRNA | BC122950.1 | - | 2 - 3 | 1153 | 97 | |
More Information
Set Membership: |
|

fam222a
Genomic Map