Assay Details
| Gene Symbol: c19orf66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097193.1 | NP_001090662.1 | 3 - 4 | 326 | 97 | |
| GenBank mRNA | BC127301.1 | - | 3 - 4 | 326 | 97 | |
More Information
Set Membership: |
|

c19orf66
Genomic Map