Assay Details
| Gene Symbol: znf268 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097271.1 | NP_001090740.1 | 590 | 95 | ||
| GenBank mRNA | BC129015.1 | - | 590 | 95 | ||
| BC166375.1 | - | 176 | 95 | |||
More Information
Set Membership: |
|

znf268
Genomic Map