Assay Details
| Gene Symbol: st8sia5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097279.1 | NP_001090748.1 | 2 - 3 | 228 | 88 | |
| XM_012956876.2 | XP_012812330.1 | 3 - 4 | 868 | 88 | ||
| XM_012956917.2 | XP_012812371.1 | 2 - 3 | 760 | 88 | ||
| GenBank mRNA | AM422136.1 | - | 2 - 3 | 228 | 88 | |
More Information
Set Membership: |
|

st8sia5
Genomic Map