Assay Details
Gene Symbol: ogfrl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097287.1 | NP_001090756.1 | 5 - 6 | 603 | 98 | |
XM_012962530.2 | XP_012817984.1 | 5 - 6 | 903 | 98 | ||
XM_018093724.1 | XP_017949213.1 | 5 - 6 | 404 | 98 | ||
GenBank mRNA | BC128651.1 | - | 5 - 6 | 603 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
