Assay Details
| Gene Symbol: b3gat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097340.1 | NP_001090809.1 | 5 - 6 | 1373 | 59 | |
| XM_012965833.2 | XP_012821287.1 | 5 - 6 | 2286 | 59 | ||
| XM_012965834.2 | XP_012821288.1 | 4 - 5 | 2122 | 59 | ||
| GenBank mRNA | BC135247.1 | - | 5 - 6 | 1373 | 59 | |
More Information
Set Membership: |
|

b3gat1
Genomic Map