Assay Details
Gene Symbol: cks1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100215.1 | NP_001093685.1 | 2 - 3 | 189 | 59 | |
GenBank mRNA | BC135373.1 | - | 2 - 3 | 189 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
