Assay Details
Gene Symbol: efnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100239.1 | NP_001093709.1 | 1 - 2 | 594 | 100 | |
GenBank mRNA | BC135636.1 | - | 1 - 2 | 594 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
