Assay Details
Gene Symbol: smn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100240.1 | NP_001093710.1 | 1 - 2 | 103 | 87 | |
XM_018092087.1 | XP_017947576.1 | 1 - 2 | 122 | 87 | ||
GenBank mRNA | BC135221.1 | - | 1 - 2 | 103 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
