Assay Details
Gene Symbol: smn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100240.1 | NP_001093710.1 | 4 - 5 | 504 | 94 | |
XM_018092087.1 | XP_017947576.1 | 3 - 4 | 397 | 94 | ||
GenBank mRNA | BC135221.1 | - | 4 - 5 | 504 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
