Assay Details
Gene Symbol: rab41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102751.1 | NP_001096221.1 | 2 - 3 | 192 | 88 | |
XM_012967863.2 | XP_012823317.1 | 2 - 3 | 218 | 88 | ||
GenBank mRNA | BC121383.1 | - | 2 - 3 | 192 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
