Assay Details
Gene Symbol: rab41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102751.1 | NP_001096221.1 | 3 - 4 | 250 | 96 | |
GenBank mRNA | BC121383.1 | - | 3 - 4 | 250 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
