Assay Details
Gene Symbol: rab41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102751.1 | NP_001096221.1 | 6 - 7 | 554 | 86 | |
XM_012967863.2 | XP_012823317.1 | 6 - 7 | 583 | 86 | ||
GenBank mRNA | BC121383.1 | - | 6 - 7 | 554 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
