Assay Details
| Gene Symbol: evc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102764.1 | NP_001096234.1 | 9 - 10 | 1461 | 68 | |
| XM_018093045.1 | XP_017948534.1 | 9 - 10 | 1498 | 68 | ||
| XM_018093058.1 | XP_017948547.1 | 9 - 10 | 1498 | 68 | ||
| GenBank mRNA | BC135352.1 | - | 9 - 10 | 1461 | 68 | |
More Information
Set Membership: |
|

evc
Genomic Map