Assay Details
| Gene Symbol: ndufa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102777.1 | NP_001096247.1 | 2 - 3 | 227 | 99 | |
| GenBank mRNA | BC135400.1 | - | 2 - 3 | 227 | 99 | |
| CT025289.2 | - | 2 - 3 | 286 | 99 | ||
| Gene Symbol: LOC100495196 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002944958.3 | XP_002945004.2 | 2 - 3 | 141 | 99 | |
More Information
Set Membership: |
|

ndufa2
Genomic Map