Assay Details
| Gene Symbol: lpar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102843.1 | NP_001096313.1 | 4 - 5 | 1015 | 69 | |
| XM_012963745.2 | XP_012819199.1 | 3 - 4 | 1143 | 69 | ||
| XM_012963778.2 | XP_012819232.1 | 3 - 4 | 1402 | 69 | ||
| GenBank mRNA | BC135679.1 | - | 4 - 5 | 1015 | 69 | |
| CT025354.2 | - | 3 - 4 | 1146 | 69 | ||
More Information
Set Membership: |
|

lpar1
Genomic Map