Assay Details
| Gene Symbol: lmcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102853.1 | NP_001096323.1 | 2 - 3 | 173 | 80 | |
| XM_012960465.2 | XP_012815919.1 | 2 - 3 | 199 | 80 | ||
| GenBank mRNA | BC135721.1 | - | 2 - 3 | 173 | 80 | |
More Information
Set Membership: |
|

lmcd1
Genomic Map