Assay Details
| Gene Symbol: arhgef9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102956.2 | NP_001096426.2 | 6 - 7 | 1150 | 63 | |
| XM_012965915.1 | XP_012821369.1 | 8 - 9 | 1768 | 63 | ||
| XM_012965916.1 | XP_012821370.1 | 6 - 7 | 1150 | 63 | ||
| XM_012965917.2 | XP_012821371.1 | 6 - 7 | 1141 | 63 | ||
| XM_012965918.2 | XP_012821372.1 | 6 - 7 | 1702 | 63 | ||
| GenBank mRNA | BC136098.1 | - | 6 - 7 | 1109 | 63 | |
More Information
Set Membership: |
|

arhgef9
Genomic Map