Assay Details
| Gene Symbol: lhfpl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012960480.2 | XP_012815934.1 | 1 - 2 | 931 | 66 | |
| GenBank mRNA | BC136166.1 | - | 1 - 2 | 839 | 66 | |
More Information
Set Membership: |
|

lhfpl4
Genomic Map