Assay Details
| Gene Symbol: farsa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079359.1 | NP_001072827.1 | 4 - 5 | 529 | 67 | |
| XM_012953178.2 | XP_012808632.1 | 3 - 4 | 747 | 67 | ||
| XM_012953179.2 | XP_012808633.1 | 4 - 5 | 550 | 67 | ||
| GenBank mRNA | BC121853.1 | - | 4 - 5 | 529 | 67 | |
| CR942740.1 | - | 4 - 5 | 510 | 67 | ||
More Information
Set Membership: |
|

farsa
Genomic Map