Assay Details
| Gene Symbol: chmp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103071.1 | NP_001096541.1 | 6 - 7 | 525 | 94 | |
| XM_012971507.2 | XP_012826961.1 | 6 - 7 | 536 | 94 | ||
| GenBank mRNA | BC135970.1 | - | 6 - 7 | 525 | 94 | |
| CT025394.2 | - | 6 - 7 | 537 | 94 | ||
More Information
Set Membership: |
|

chmp6
Genomic Map