Assay Details
| Gene Symbol: slc41a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001348526.1 | NP_001335455.1 | 2 - 3 | 654 | 77 | |
| GenBank mRNA | BC167928.1 | - | 2 - 3 | 614 | 77 | |
| CX807405.1 | - | 2 - 3 | 654 | 77 | ||
More Information
Set Membership: |
|

slc41a2
Genomic Map