Assay Details
| Gene Symbol: vamp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112930.2 | NP_001106401.2 | 3 - 4 | 281 | 70 | |
| GenBank mRNA | BC152063.1 | - | 3 - 4 | 244 | 70 | |
More Information
Set Membership: |
|

vamp1
Genomic Map