Assay Details
| Gene Symbol: negr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113066.1 | NP_001106537.1 | 1 - 2 | 341 | 67 | |
| XM_012960537.2 | XP_012815991.1 | 1 - 2 | 1206 | 67 | ||
| GenBank mRNA | BC154906.1 | - | 1 - 2 | 341 | 67 | |
More Information
Set Membership: |
|

negr1
Genomic Map