Assay Details
| Gene Symbol: mvb12a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113116.1 | NP_001106587.1 | 6 - 7 | 590 | 66 | |
| XM_018096132.1 | XP_017951621.1 | 6 - 7 | 966 | 66 | ||
| GenBank mRNA | BC155409.1 | - | 6 - 7 | 590 | 66 | |
More Information
Set Membership: |
|

mvb12a
Genomic Map